Imprimer
Human Syndecan-1 ELISA Kits (CD138)
Marques : Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB46506-96 AB46506-2X96
ABCAAB46506-96EA 969 EUR
ABCAAB46506-96 ABCAAB46506-2X96
Human Syndecan-1 ELISA Kits (CD138)
Tests Tests ELISA
Human Syndecan-1 ELISA Kit (CD138) is a Sandwich ELISA used to quantify Human Syndecan-1 (CD138) with a sensitivity of 4,94 ng/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Wide dynamic range – 8 to 256 ng/ml - Cited in over 30 citations.