Imprimer
Rat IL-4 ELISA Set (without plates)
Marques : Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB48423-480 AB48423-15X96 AB48423-10X96
ABCAAB48423-480EA 1140 EUR
ABCAAB48423-480 ABCAAB48423-15X96 ABCAAB48423-10X96
Rat IL-4 ELISA Set (without plates)
Tests Tests ELISA
Rat IL-4 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Rat IL-4 ELISA Set (without plates) in Rat in plasma, cell culture supernatant, serum samples.