Imprimer
Human IFN gamma High Sensitivity ELISA Kit
Marques : Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB46048-96 AB46048-2X96
ABCAAB46048-96EA 1010 EUR
ABCAAB46048-96 ABCAAB46048-2X96
Human IFN gamma High Sensitivity ELISA Kit
Tests Tests ELISA
Human IFN gamma High Sensitivity ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IFN gamma High Sensitivity in Human in Plasma, Cell culture supernatant, Serum samples.