Human CTHRC1 ELISA Kit
Marques : AbcamTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Human CTHRC1 ELISA kit is a single-wash 90-min Simplestep used to quantify Human CTHRC1 with a sensitivity of 27,3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair.