Imprimer
rubipor XS Safety Gloves, 60316 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6031606 6031608 6031609 6031610 6031607
UVEX6031606EA 55.6 EUR
UVEX6031606 UVEX6031608 UVEX6031609 UVEX6031610 UVEX6031607
rubipor XS Safety Gloves, 60316 Series
Gants
The rubipor XS is a lightweight, elastic safety glove with breathable NBR impregnation.
  • Lightweight, elastic safety glove with stretch cotton material
  • Palm and fingertips coated with breathable special NBR impregnation
  • Suitable for operating touch displays
The combination of the very thin layer of impregnation and the stretch cotton material provides an extremely good level of breathability. Free of harmful substances in line with OEKO-Tex® Standard 100.