Imprimer
Top Grade 8300 Leather Safety Gloves, 60292 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6029209 6029210 6029211
UVEX6029209EA 7 EUR
UVEX6029209 UVEX6029210 UVEX6029211
Top Grade 8300 Leather Safety Gloves, 60292 Series
Gants
An extremely high quality leather safety glove for mechanical applications.
  • Soft, supple leather
  • Exceptional abrasion resistance and cut resistance properties
  • Excellent wearer comfort
The uvex top grade series is characterised by very high-quality leather. Various sizes and consistently high quality enable particularly good wearer comfort and a long service life.