Imprimer
HexArmor® Rig Lizard® 2021X Safety Gloves, 60670 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6067006 6067010 6067007 6067008 6067009 6067011 6067012
UVEX6067006EA 0 EUR
UVEX6067006 UVEX6067010 UVEX6067007 UVEX6067008 UVEX6067009 UVEX6067011 UVEX6067012
HexArmor® Rig Lizard® 2021X Safety Gloves, 60670 Series
Gants
The Rig Lizard® 2021X is a premium hi-vis safety glove with a sewn-on IR-X® Impact Exoskeleton and a durable TP-X® palm with reinforced stitching for high-performing protection.
  • IR-X® Impact Exoskeleton™ with high-flex design for sewn-on back-of-hand impact resistance
  • High durability for long-lasting protection
  • Hi-vis colour scheme
Offering excellent durability in cut protection level C, this comfortable, high-performance glove provides optimal dexterity and tactility. Back-of-hand IR-X® Impact Exoskeleton™ keeps hands safe from smash and impact hazards. Durable TP-X® palm with reinforced stitching provides superior wet/dry grip and abrasion resistance. Reinforced index finger and thumb saddle ensures increased durability. An elastic cuff provides a secure fit.