Imprimer
HexArmor® Chrome 4026, Safety Gloves, 60986 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6098606 6098607 6098608 6098609 6098610 6098611 6098612
UVEX6098606EA 0 EUR
UVEX6098606 UVEX6098607 UVEX6098608 UVEX6098609 UVEX6098610 UVEX6098611 UVEX6098612
HexArmor® Chrome 4026, Safety Gloves, 60986 Series
Gants
The Chrome Series® 4026 safety gloves feature a Mud Grip® palm that enhances grip in dry and light oil applications, impact protection, and 360° cut resistance for high-performing protection on the job.
  • High cut resistance with CE 4X21FP
  • SuperFabric® material protects against hazards
  • Durable Mud Grip® palm
  • Elastic cuff with Velcro®
  • Form-fitting design
  • Launderable
Featuring a form-fitting design for increased dexterity and comfort, these gloves include SuperFabric® material to protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. Back-of-hand impact protection ensures high durability for long-lasting protection. Durable Mud Grip® palm provides the ultimate durability and grip in dry or light oil applications. Elastic cuff with Velcro® closure allows for a comfortable and precise fit.