Imprimer
phynomic allround Safety Gloves, 60049 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6004905 6004912
UVEX6004905EA 66.2 EUR
UVEX6004905 UVEX6004912
phynomic allround Safety Gloves, 60049 Series
Gants
The uvex phynomic allround safety glove is a versatile choice for various mechanical tasks, offering a perfect balance of protection and comfort.
  • Suitable for use in a range of fields
  • Dirt-resistant
  • Excellent skin tolerance and free from pollutants
Featuring an aqua-polymer foam coating on the palm and fingertips, these gloves provide excellent grip and mechanical abrasion resistance, even in slightly damp conditions. The lightweight and flexible design ensures a snug fit, making them feel like a second skin. With high breathability and outstanding tactile sensitivity, these gloves are ideal for precision work. Additionally, they are dermatologically tested and free from harmful substances, ensuring excellent skin compatibility.