Imprimer
C300 dry Cut Protection Gloves, 60549 Series
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6054907 6054908 6054909 6054910 6054911
UVEX6054907EA 147 EUR
UVEX6054907 UVEX6054908 UVEX6054909 UVEX6054910 UVEX6054911
C300 dry Cut Protection Gloves, 60549 Series
Gants
The C300 cut protection glove range sets standards for protection, comfort, tactility, and efficiency. These innovative, hi-tech protective gloves combine all of these qualities, with level 3/C cut protection.
  • Highly flexible
  • Very good tactile sensation
  • Perfect fit with 3D Ergo technology
  • Outstanding wearer comfort due to uvex climazone
This cut protection glove with excellent wearer comfort features palm and fingers with a High-Performance Vinyl (HPV) grip. Free of harmful substances in accordance with OEKO-TEX® Standard 100.