Imprimer

Anti-Factor IX/PTC Rabbit Monoclonal Antibody [clone: EPR28007-7]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB320865-100UG AB320865-1MG
ABCAAB320865-100UGEA 919 EUR
ABCAAB320865-100UG ABCAAB320865-1MG
Anti-Factor IX/PTC Rabbit Monoclonal Antibody [clone: EPR28007-7]
Anticorps
Rabbit monoclonal [EPR28007-7] to Factor IX/PTC - BSA and Azide free (Capture).

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Factor IX/PTC
Clonality: Monoclonal
Clone: EPR28007-7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR