Imprimer

Anti-Metapneumovirus Fusion Protein Rabbit Monoclonal Antibody [clone: EPR29365-579]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB320652-20UL AB320652-100UL AB320652-1ML
ABCAAB320652-20ULEA 244 EUR
ABCAAB320652-20UL ABCAAB320652-100UL ABCAAB320652-1ML
Anti-Metapneumovirus Fusion Protein Rabbit Monoclonal Antibody [clone: EPR29365-579]
Anticorps
Rabbit monoclonal [EPR29365-579] to Metapneumovirus Fusion Protein.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Metapneumovirus Fusion Protein
Clonality: Monoclonal
Clone: EPR29365-579
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR