Imprimer

Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR28045-139]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB315164-20UL AB315164-100UL AB315164-1ML
ABCAAB315164-20ULEA 244 EUR
ABCAAB315164-20UL ABCAAB315164-100UL ABCAAB315164-1ML
Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR28045-139]
Anticorps
Rabbit monoclonal [EPR28045-139] to AIRE.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AIRE
Clonality: Monoclonal
Clone: EPR28045-139
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR