Imprimer

Anti-CD62E Rabbit Monoclonal Antibody [clone: EPR25616-62] (Alexa Fluor® 488)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB318169-10UL AB318169-100UL
ABCAAB318169-10ULEA 238 EUR
ABCAAB318169-10UL ABCAAB318169-100UL
Anti-CD62E Rabbit Monoclonal Antibody [clone: EPR25616-62] (Alexa Fluor® 488)
Anticorps
Alexa Fluor® 488 Rabbit monoclonal [EPR25616-62] to CD62E.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD62E
Clonality: Monoclonal
Clone: EPR25616-62
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR