Imprimer
Anti-LONP1/Lon Rabbit Monoclonal Antibody [clone: EPR28510-62]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB317561-20UL AB317561-100UL AB317561-1ML
ABCAAB317561-20ULEA 244 EUR
ABCAAB317561-20UL ABCAAB317561-100UL ABCAAB317561-1ML
Anti-LONP1/Lon Rabbit Monoclonal Antibody [clone: EPR28510-62]
Anticorps
Rabbit monoclonal [EPR28510-62] to LONP1/Lon.
Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LONP1/Lon
Clonality: Monoclonal
Clone: EPR28510-62
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat