Imprimer

Anti-Histone H3 (citrulline R2 + R8 + R17) Rabbit Recombinant Multiclonal Antibody [clone: RM1001]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB283863-100 AB283863-1
ABCAAB283863-100EA 1020 EUR
ABCAAB283863-100 ABCAAB283863-1
Anti-Histone H3 (citrulline R2 + R8 + R17) Rabbit Recombinant Multiclonal Antibody [clone: RM1001]
Anticorps
Rabbit recombinant multiclonal [RM1001] to Histone H3 (citrulline R2 + R8 + R17) - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Recombinant Multiclonal
Clone: RM1001
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR