Imprimer

Anti-Cleaved PARP1 Rabbit Monoclonal Antibody [clone: SP276]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB225715-100 AB225715-1 AB225715-500UL AB225715-20UL
ABCAAB225715-100EA 475 EUR
ABCAAB225715-100 ABCAAB225715-1 ABCAAB225715-500UL ABCAAB225715-20UL
Anti-Cleaved PARP1 Rabbit Monoclonal Antibody [clone: SP276]
Anticorps
Rabbit monoclonal [SP276] to Cleaved PARP1.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cleaved PARP1
Clonality: Monoclonal
Clone: SP276
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR