Imprimer

Anti-PI 3 Kinase catalytic subunit gamma/PI3K-gamma Rabbit Monoclonal Antibody [clone: EPR25156-60]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB302958-100 AB302958-20UL AB302958-1ML
ABCAAB302958-100EA 707 EUR
ABCAAB302958-100 ABCAAB302958-20UL ABCAAB302958-1ML
Anti-PI 3 Kinase catalytic subunit gamma/PI3K-gamma Rabbit Monoclonal Antibody [clone: EPR25156-60]
Anticorps
Rabbit monoclonal [EPR25156-60] to PI 3 Kinase catalytic subunit gamma/PI3K-gamma.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase catalytic subunit gamma/PI3K-gamma
Clonality: Monoclonal
Clone: EPR25156-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR