Imprimer

Anti-PI 3 Kinase catalytic subunit alpha/PIK3CA Rabbit Monoclonal Antibody [clone: EP383Y]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB185927-100 AB185927-1
ABCAAB185927-100EA 1020 EUR
ABCAAB185927-100 ABCAAB185927-1
Anti-PI 3 Kinase catalytic subunit alpha/PIK3CA Rabbit Monoclonal Antibody [clone: EP383Y]
Anticorps
Rabbit monoclonal [EP383Y] to PI 3 Kinase catalytic subunit alpha/PIK3CA - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase catalytic subunit alpha/PIK3CA
Clonality: Monoclonal
Clone: EP383Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR