Imprimer

Anti-Dopamine beta Hydroxylase Rabbit Monoclonal Antibody [clone: EPR23024-304]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB277245-100 AB277245-1
ABCAAB277245-100EA 869 EUR
ABCAAB277245-100 ABCAAB277245-1
Anti-Dopamine beta Hydroxylase Rabbit Monoclonal Antibody [clone: EPR23024-304]
Anticorps
Rabbit monoclonal [EPR23024-304] to Dopamine beta Hydroxylase - BSA and Azide free (Capture).

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Dopamine beta Hydroxylase
Clonality: Monoclonal
Clone: EPR23024-304
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR