Imprimer
Anti-DNAJC5B Rabbit Monoclonal Antibody [clone: EPR24152-62]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB281840-100 AB281840-1
ABCAAB281840-100EA 1020 EUR
ABCAAB281840-100 ABCAAB281840-1
Anti-DNAJC5B Rabbit Monoclonal Antibody [clone: EPR24152-62]
Anticorps
Rabbit monoclonal [EPR24152-62] to DNAJC5B - BSA and Azide free.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: DNAJC5B
Clonality: Monoclonal
Clone: EPR24152-62
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat