Imprimer
Anti-Histone H2B Rabbit Monoclonal Antibody [clone: EPR27298-41]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB313789-100UL AB313789-20UL AB313789-1ML
ABCAAB313789-100ULEA 694 EUR
ABCAAB313789-100UL ABCAAB313789-20UL ABCAAB313789-1ML
Anti-Histone H2B Rabbit Monoclonal Antibody [clone: EPR27298-41]
Anticorps
Rabbit monoclonal [EPR27298-41] to Histone H2B (mutated E76K).
Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EPR27298-41
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human