Imprimer

Anti-PI 3 Kinase Class 2A/Cpk Mouse Monoclonal Antibody [clone: 17/PI3-Kinase p170]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB307099-100 AB307099-1
ABCAAB307099-100EA 1020 EUR
ABCAAB307099-100 ABCAAB307099-1
Anti-PI 3 Kinase Class 2A/Cpk Mouse Monoclonal Antibody [clone: 17/PI3-Kinase p170]
Anticorps
Mouse monoclonal [17/PI3-Kinase p170] to PI 3 Kinase Class 2A/Cpk - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase Class 2A/Cpk
Clonality: Monoclonal
Clone: 17/PI3-Kinase p170
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR