Imprimer

Anti-AKR1B10 Rabbit Monoclonal Antibody [clone: EPR22343-275]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB259724-100 AB259724-1
ABCAAB259724-100EA 938 EUR
ABCAAB259724-100 ABCAAB259724-1
Anti-AKR1B10 Rabbit Monoclonal Antibody [clone: EPR22343-275]
Anticorps
Rabbit monoclonal [EPR22343-275] to AKR1B10 - BSA and Azide free (Detector).

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AKR1B10
Clonality: Monoclonal
Clone: EPR22343-275
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR