Imprimer
Anti-NFAT1 Mouse Monoclonal Antibody [clone: 1/NFAT-1]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB288768-100 AB288768-1MG AB288768-20UG
ABCAAB288768-100EA 707 EUR
ABCAAB288768-100 ABCAAB288768-1MG ABCAAB288768-20UG
Anti-NFAT1 Mouse Monoclonal Antibody [clone: 1/NFAT-1]
Anticorps
Mouse monoclonal [1/NFAT-1] to NFAT1.
Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NFAT1
Clonality: Monoclonal
Clone: 1/NFAT-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human