Imprimer
Anti-CD1c Rabbit Monoclonal Antibody [clone: EPR23189-305] (Alexa Fluor® 488)
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB305271-10 AB305271-100
ABCAAB305271-10EA 225 EUR
ABCAAB305271-10 ABCAAB305271-100
Anti-CD1c Rabbit Monoclonal Antibody [clone: EPR23189-305] (Alexa Fluor® 488)
Anticorps
Alexa Fluor® 488 Rabbit monoclonal [EPR23189-305] to CD1c.
Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD1c
Clonality: Monoclonal
Clone: EPR23189-305
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human