Imprimer

Anti-Junctional Adhesion Molecule 2/JAM-B Rabbit Monoclonal Antibody [clone: EPR2489(2)]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB156586-100 AB156586-1ML
ABCAAB156586-100EA 782 EUR
ABCAAB156586-100 ABCAAB156586-1ML
Anti-Junctional Adhesion Molecule 2/JAM-B Rabbit Monoclonal Antibody [clone: EPR2489(2)]
Anticorps
Rabbit monoclonal [EPR2489(2)] to Junctional Adhesion Molecule 2/JAM-B.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Junctional Adhesion Molecule 2/JAM-B
Clonality: Monoclonal
Clone: EPR2489(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR