Imprimer

Anti-Histone H2B (crotonyl K5) Rabbit Monoclonal Antibody [clone: EPR17483]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB177396-100 AB177396-1ML
ABCAAB177396-100EA 650 EUR
ABCAAB177396-100 ABCAAB177396-1ML
Anti-Histone H2B (crotonyl K5) Rabbit Monoclonal Antibody [clone: EPR17483]
Anticorps
Rabbit monoclonal [EPR17483] to Histone H2B (crotonyl K5).

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EPR17483
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR