Imprimer

Anti-Eph receptor A3 Rabbit Monoclonal Antibody [clone: EPR24479-136]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB281926-100 AB281926-20UL AB281926-1ML
ABCAAB281926-100EA 663 EUR
ABCAAB281926-100 ABCAAB281926-20UL ABCAAB281926-1ML
Anti-Eph receptor A3 Rabbit Monoclonal Antibody [clone: EPR24479-136]
Anticorps
Rabbit monoclonal [EPR24479-136] to Eph receptor A3.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Eph receptor A3
Clonality: Monoclonal
Clone: EPR24479-136
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR