FITC Rabbit monoclonal [PI3KY458-1A11] to PI 3 Kinase p85 alpha (phospho Y467) + PI3 Kinase p55 (phospho Y199).
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PI 3 Kinase p85 alpha+PI3 Kinase p55
Clonality: Monoclonal
Clone: PI3KY458-1A11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human