Imprimer

Anti-PODXL Mouse Monoclonal Antibody [clone: PcMab-47]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB264549-100 AB264549-1
ABCAAB264549-100EA 1020 EUR
ABCAAB264549-100 ABCAAB264549-1
Anti-PODXL Mouse Monoclonal Antibody [clone: PcMab-47]
Anticorps
Mouse monoclonal [PcMab-47] to PODXL - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: PcMab-47
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR