Imprimer

Anti-Histone H3 (tri methyl K9) Rabbit Monoclonal Antibody [clone: EPR16601]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB232324-100 AB232324-1
ABCAAB232324-100EA 1020 EUR
ABCAAB232324-100 ABCAAB232324-1
Anti-Histone H3 (tri methyl K9) Rabbit Monoclonal Antibody [clone: EPR16601]
Anticorps
Rabbit monoclonal [EPR16601] to Histone H3 (tri methyl K9) - ChIP Grade – BSA and Azide free.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR16601
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR