Imprimer

Anti-MASH1/Achaete-scute homolog 1 Rabbit Monoclonal Antibody [clone: EPR19840]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB240385-1 AB240385-100 AB240385-10
ABCAAB240385-1EA 9160 EUR
ABCAAB240385-1 ABCAAB240385-100 ABCAAB240385-10
Anti-MASH1/Achaete-scute homolog 1 Rabbit Monoclonal Antibody [clone: EPR19840]
Anticorps
Rabbit monoclonal [EPR19840] to MASH1/Achaete-scute homolog 1 - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MASH1/Achaete-scute homolog 1
Clonality: Monoclonal
Clone: EPR19840
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR