Imprimer

Anti-P2Y12 Rabbit Monoclonal Antibody [clone: EPR26298-93] (Alexa Fluor® 488)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB309607-100UL AB309607-10UL
ABCAAB309607-100ULEA 819 EUR
ABCAAB309607-100UL ABCAAB309607-10UL
Anti-P2Y12 Rabbit Monoclonal Antibody [clone: EPR26298-93] (Alexa Fluor® 488)
Anticorps
Alexa Fluor® 488 Rabbit monoclonal [EPR26298-93] to P2Y12.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: P2Y12
Clonality: Monoclonal
Clone: EPR26298-93
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR