Imprimer

Anti-PMCA1 Rabbit Monoclonal Antibody [clone: EPR12029]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB190355-100 AB190355-1ML
ABCAAB190355-100EA 669 EUR
ABCAAB190355-100 ABCAAB190355-1ML
Anti-PMCA1 Rabbit Monoclonal Antibody [clone: EPR12029]
Anticorps
Rabbit monoclonal [EPR12029] to PMCA1.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PMCA1
Clonality: Monoclonal
Clone: EPR12029
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR