Imprimer

Anti-LILRB3 Rat Monoclonal Antibody [clone: FRAS92B]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB271287-100 AB271287-1MG
ABCAAB271287-100EA 732 EUR
ABCAAB271287-100 ABCAAB271287-1MG
Anti-LILRB3 Rat Monoclonal Antibody [clone: FRAS92B]
Anticorps
Rat monoclonal [FRAS92B] to LILRB3.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LILRB3
Clonality: Monoclonal
Clone: FRAS92B
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR