Imprimer

Anti-Somatostatin Receptor 2 Rabbit Monoclonal Antibody [clone: UMB1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB216649-100 AB216649-1
ABCAAB216649-100EA 1020 EUR
ABCAAB216649-100 ABCAAB216649-1
Anti-Somatostatin Receptor 2 Rabbit Monoclonal Antibody [clone: UMB1]
Anticorps
Rabbit monoclonal [UMB1] to Somatostatin Receptor 2 - Low endotoxin, Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Somatostatin Receptor 2
Clonality: Monoclonal
Clone: UMB1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR