Imprimer

Anti-LAMP1 Mouse Monoclonal Antibody [clone: 25/Lamp-1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB289548-100 AB289548-1MG AB289548-20UG
ABCAAB289548-100EA 707 EUR
ABCAAB289548-100 ABCAAB289548-1MG ABCAAB289548-20UG
Anti-LAMP1 Mouse Monoclonal Antibody [clone: 25/Lamp-1]
Anticorps
Mouse monoclonal [25/Lamp-1] to LAMP1.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 25/Lamp-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR