Imprimer

Anti-Scavenging Receptor SR-BI Rabbit Monoclonal Antibody [clone: EPR20190]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB217318-100 AB217318-1ML
ABCAAB217318-100EA 700 EUR
ABCAAB217318-100 ABCAAB217318-1ML
Anti-Scavenging Receptor SR-BI Rabbit Monoclonal Antibody [clone: EPR20190]
Anticorps
Rabbit monoclonal [EPR20190] to Scavenging Receptor SR-BI.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Scavenging Receptor SR-BI
Clonality: Monoclonal
Clone: EPR20190
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR