Imprimer
Anti-PDE4A Rabbit Monoclonal Antibody [clone: EPR26991-62]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB313720-1MG AB313720-100UG
ABCAAB313720-1MGEA 9200 EUR
ABCAAB313720-1MG ABCAAB313720-100UG
Anti-PDE4A Rabbit Monoclonal Antibody [clone: EPR26991-62]
Anticorps
Rabbit monoclonal [EPR26991-62] to PDE4A - BSA and Azide free (Detector).
Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PDE4A
Clonality: Monoclonal
Clone: EPR26991-62
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human