Imprimer
Filtration funnels, cylindrical
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3770/06 3770/02 3770/04 3770/08 3770/10
BIBB3770/06EA 578 EUR
BIBB3770/06 BIBB3770/02 BIBB3770/04 BIBB3770/08 BIBB3770/10
Filtration funnels, cylindrical
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Made of PYREX® borosilicate glass, these are ideal for filtering precipitates.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.