Imprimer
Filtration funnels
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3730/06 3730/08 3750/03
BIBB3730/06EA 224 EUR
BIBB3730/06 BIBB3730/08 BIBB3750/03
Filtration funnels
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Pyrex® borosilicate glass.
  • Sintered glass disc
  • Filter paper is not required for filtration of solutions
  • Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid