Imprimer
Cryoboxes
Fournisseur: SARSTEDT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
95.64.922 95.64.923 95.64.924 95.64.981 95.64.982 95.64.997
SARS95.64.922EA 1580 EUR
SARS95.64.922 SARS95.64.923 SARS95.64.924 SARS95.64.981 SARS95.64.982 SARS95.64.997
Cryoboxes
Boîtes Boîtes pour cryotubes
Storage boxes, moisture-resistant laminated carboard material with lid and compartmentalised insert.
  • Space saving and organised sample storage
  • Safe and light-protected storage
Suitable for micro tubes, PCR tubes and centrifuge tubes.