Imprimer
Serological pipettes
Fournisseur: SARSTEDT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
86.1252.025 86.1254.025 86.1254.001 86.1251.001 86.1253.001 86.1685.001 86.1251.025 86.1252.001 86.1253.025 86.1685.020
SARS86.1252.025EA 356 EUR
SARS86.1252.025 SARS86.1254.025 SARS86.1254.001 SARS86.1251.001 SARS86.1253.001 SARS86.1685.001 SARS86.1251.025 SARS86.1252.001 SARS86.1253.025 SARS86.1685.020
Serological pipettes
Pipettes Pipettes sérologiques
PS, graduated, sterile.
  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging