Imprimer
Microcentrifuge tubes, with or without attached caps
Fournisseur: SARSTEDT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72.690 72.690.007 72.690.475 72.690.477 72.690.478 72.696 72.708 72.688 72.690.001 72.690.550 72.709 72.691
SARS72.690EA 276 EUR
SARS72.690 SARS72.690.007 SARS72.690.475 SARS72.690.477 SARS72.690.478 SARS72.696 SARS72.708 SARS72.688 SARS72.690.001 SARS72.690.550 SARS72.709 SARS72.691
Microcentrifuge tubes, with or without attached caps
Tubes Microtubes à centrifuger
PP, translucent.
  • Autoclavable
  • Good chemical resistance
  • Non sterile
For transport, storage and sample processing in the laboratory.