Imprimer
Adapters with stopcock and socket
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MF14/2/SC MF14/3/SC
201-3019EA 117 EUR
201-3019 BIBBMF14/3/SC
Adapters with stopcock and socket
Adaptateurs Raccords rodés
PYREX®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .