Imprimer
PYREX® Bottles, Media-lab, Clear, Square Shape With Cap and Pouring Ring
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1520/08D 1520/10D 1520/04D 1520/06D
BIBB1520/08DEA 147 EUR
BIBB1520/08D BIBB1520/10D BIBB1520/04D BIBB1520/06D
PYREX® Bottles, Media-lab, Clear, Square Shape With Cap and Pouring Ring
Flacons Flacons en verre
The innovative square shape of these glass bottles allows them to be stacked closer together than traditional round bottles.
  • Square shape allows closer stacking in water baths or on shelves
  • Up to 20% saving of space compared to round bottles
  • Bottles supplied complete with polypropylene cap and pouring ring
  • For use up to 140 °C
Printed with Trace Code for downloadable batch certificate.