Imprimer
Flacons PYREX®, Media-Lab, ambrés, sans bouchon ni bague anti-goutte, DWK Life Sciences
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1519/06 1519/08 1519/10 1519/01 1519/02 1519/04 1519/12 1519/18 1519/16
BIBB1519/06EA 417 EUR
BIBB1519/06 BIBB1519/08 BIBB1519/10 BIBB1519/01 BIBB1519/02 BIBB1519/04 BIBB1519/12 BIBB1519/18 BIBB1519/16
Flacons PYREX®, Media-Lab, ambrés, sans bouchon ni bague anti-goutte, DWK Life Sciences
Flacons Flacons en verre
Ideal for the storage of light sensitive materials, these bottles are amber coated versions of the iconic GL 45 screw threaded media-lab bottles. Supplied without the blue polypropylene screw cap and clear pouring ring.
  • Bottles only; supplied without cap or pouring ring
  • Manufactured from amber-coated borosilicate glass
  • UV absorbent up to 500 nm
  • Ideal for storage of light sensitive materials
  • Printed with trace code for downloadable batch certificate
Never autoclave bottles with a fully tightened screw cap. Always slacken off. Failure to follow this procedure can lead to breakage of the bottle.
The 25 ml size bottle features a glass lip and does not require a pouring ring.
Certifications:
Complies with ISO 4796.