Imprimer
Fernbach culture flasks
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1138/10 1139/10
391-0533EA 513 EUR
391-0533 391-0534
Fernbach culture flasks
Ballons Flacons de culture
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
  • Exhibit 3 baffle indents to provide optimal and consistent agitation
  • Large surface area to volume ratio
  • Available in two versions, with Delong neck or screw cap