Imprimer
Stopcocks, three way, T-bore
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SST304 SST3025
BIBBSST304EA 561 EUR
BIBBSST304 BIBBSST3025
Stopcocks, three way, T-bore
Robinets d'arrêt
Pyrex® borosilicate glass with PTFE key
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable